Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl imidazo[1,2-b]pyridazine-2-carboxylate 95%

Supplier: AMBEED, INC
Warning

A479793-100MG A479793-1G A479793-250MG
77156-436EA 13 USD InStock
77156-436 77156-438 77156-440
Ethyl imidazo[1,2-b]pyridazine-2-carboxylate 95%
Ethyl imidazo[1,2-b]pyridazine-2-carboxylate
Precaución: for Research Use Only


Formula: C₉H₉N₃O₂
MDL Number: MFCD17016047
CAS Number: 123531-27-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty