Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,5-Dimethyl-1H-pyrazole-3-carboxamide 95%

Supplier: AMBEED, INC
Warning

A486208-100MG A486208-1G A486208-250MG A486208-5G
77156-494EA 25 USD InStock
77156-494 77156-496 77156-498 77156-500
1,5-Dimethyl-1H-pyrazole-3-carboxamide 95%
1,5-Dimethyl-1H-pyrazole-3-carboxamide
Precaución: for Research Use Only


Formula: C₆H₉N₃O
MDL Number: MFCD05664820
CAS Number: 54384-74-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty