Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Phenylpiperazine dihydrochloride 95%

Supplier: AMBEED, INC
Warning

A491424-1G A491424-250MG A491424-5G
77156-566EA 17 USD InStock
77156-566 77156-568 77156-570
2-Phenylpiperazine dihydrochloride 95%
2-Phenylpiperazine dihydrochloride
Precaución: for Research Use Only


Formula: C₁₀H₁₄N₂·2ClH
MDL Number: MFCD03840168
CAS Number: 872139-22-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty