Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl 98%

Supplier: AMBEED, INC
Warning

A491448-10G A491448-1G A491448-250MG A491448-5G
77156-572EA 281 USD InStock
77156-572 77156-574 77156-576 77156-578
(S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl 98%
(S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl
Precaución: for Research Use Only


Formula: C₄₈H₄₀P₂
MDL Number: MFCD01311709
CAS Number: 100165-88-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty