Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Phenyl trifluoromethyl sulfone 98%

Supplier: AMBEED, INC
Warning

A506874-1G A506874-250MG A506874-5G A506874-25G
77156-694EA 21 USD InStock
77156-694 77156-696 77594-590 77686-320
Phenyl trifluoromethyl sulfone 98%
Phenyl trifluoromethyl sulfone
Precaución: for Research Use Only


Formula: C₇H₅F₃O₂S
MDL Number: MFCD00159083
CAS Number: 426-58-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty