Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-2-Aminohexan-1-ol hydrochloride 95%

Supplier: AMBEED, INC
Warning

A513786-100MG A513786-1G A513786-250MG
77156-796EA 5 USD InStock
77156-796 77156-798 77156-800
(R)-2-Aminohexan-1-ol hydrochloride 95%
(R)-2-Aminohexan-1-ol hydrochloride
Precaución: for Research Use Only


Formula: C₆H₁₆ClNO
MDL Number: MFCD28986146
CAS Number: 158741-04-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty