Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

BGT226 maleate 98+%

Supplier: AMBEED, INC
Warning

A514784-10MG A514784-1MG
77156-814EA 61 USD InStock
77156-814 77156-816 77156-818
BGT226 maleate 98+%
BGT226 maleate
Precaución: for Research Use Only


Formula: C₂₈H₂₅F₃N₆O₂·C₄H₄O₄
MW: 650.6 g/mol
MDL Number: MFCD22124895
CAS Number: 1245537-68-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty