Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(5-(3-(4-(2-Bromo-5-fluorophenoxy)piperidin-1-yl)isoxazol-5-yl)-2H-tetrazol-2-yl)acetic acid 98%

Supplier: AMBEED, INC
Danger

A515360-10MG A515360-1MG A515360-25MG A515360-2MG A515360-50MG A515360-5MG
77156-820EA 156 USD InStock
77156-820 77156-822 77156-824 77156-826 77156-828 77156-830
2-(5-(3-(4-(2-Bromo-5-fluorophenoxy)piperidin-1-yl)isoxazol-5-yl)-2H-tetrazol-2-yl)acetic acid 98%
2-(5-(3-(4-(2-Bromo-5-fluorophenoxy)piperidin-1-yl)isoxazol-5-yl)-2H-tetrazol-2-yl)acetic acid
Precaución: for Research Use Only


Formula: C₁₇H₁₆BrFN₆O₄
MW: 467.25 g/mol
MDL Number: MFCD20039622
CAS Number: 1030612-90-8
UN: 3077
ADR: 9 Ⅲ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty