Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-2-fluoronicotinic acid 97%

Supplier: AMBEED, INC
Warning

A520759-100MG A520759-1G A520759-250MG
77156-868EA 18 USD InStock
77156-868 77156-870 77156-872
6-Bromo-2-fluoronicotinic acid 97%
6-Bromo-2-fluoronicotinic acid
Precaución: for Research Use Only


Formula: C₆H₃BrFNO₂
MDL Number: MFCD13185804
CAS Number: 1214345-17-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty