Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

URMC-099 98+%

Supplier: AMBEED, INC
Warning

A521108-10MG A521108-1MG A521108-25MG A521108-5MG
77156-888EA 42 USD InStock
77156-888 77156-890 77156-892 77156-894
URMC-099 98+%
URMC-099
Precaución: for Research Use Only


Formula: C₂₇H₂₇N₅
MDL Number: MFCD28168077
CAS Number: 1229582-33-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty