Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl N-(carbamothioylmethyl)carbamate 98%

Supplier: AMBEED, INC
Warning

A521144-1G A521144-250MG A521144-25G A521144-5G
77156-896EA 30 USD InStock
77156-896 77156-898 77156-900 77156-902
tert-Butyl N-(carbamothioylmethyl)carbamate 98%
tert-Butyl N-(carbamothioylmethyl)carbamate
Precaución: for Research Use Only


Formula: C₇H₁₄N₂O₂S
MDL Number: MFCD09025922
CAS Number: 89226-13-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty