Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Menaquinone 7 98%

Supplier: AMBEED, INC
Warning

A532004-1MG A532004-250MG A532004-5MG A532004-1G
77156-996EA 13 USD InStock
77156-996 77156-998 77157-000 77686-373
Menaquinone 7 98%
Menaquinone 7
Precaución: for Research Use Only


Formula: C₄₆H₆₄O₂
MDL Number: MFCD06200757
CAS Number: 2124-57-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty