Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethoxytriethylsilane 99%

Supplier: AMBEED, INC
Danger

A536568-1G A536568-25G A536568-5G
77157-022EA 6 USD InStock
77157-022 77157-024 77157-026
Ethoxytriethylsilane 99%
Ethoxytriethylsilane
Precaución: for Research Use Only


Formula: C₈H₂₀OSi
MDL Number: MFCD00054718
CAS Number: 597-67-1
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty