Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Methyl-1-imidazolecarboxamide 95%

Supplier: AMBEED, INC
Warning

A536796-10G A536796-1G A536796-250MG A536796-25G A536796-5G
77157-032EA 157 USD InStock
77157-032 77157-034 77157-036 77157-038 77157-040
N-Methyl-1-imidazolecarboxamide 95%
N-Methyl-1-imidazolecarboxamide
Precaución: for Research Use Only


Formula: C₅H₇N₃O
MW: 125.12 g/mol
MDL Number: MFCD11099725
CAS Number: 72002-25-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty