Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2',6'-Difluoro-3'-methylacetophenone 98+%

Supplier: AMBEED, INC
Warning

A539134-1G A539134-250MG
77157-078EA 25 USD InStock
77157-078 77157-080
2',6'-Difluoro-3'-methylacetophenone 98+%
2',6'-Difluoro-3'-methylacetophenone
Precaución: for Research Use Only


Formula: C₉H₈F₂O
MW: 170.16
MDL Number: MFCD01631339
CAS Number: 261763-31-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty