Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α,α,α-Trifluoro-o-xylene 98%

Supplier: AMBEED, INC
Danger

A546661-1G A546661-5G
77157-118EA 28.84 USD InStock
77157-118 77157-120 77157-122
α,α,α-Trifluoro-o-xylene 98%
α,α,α-Trifluoro-o-xylene
Precaución: for Research Use Only


Formula: C₈H₇F₃
MW: 160.14
Boiling Pt: 126
Density: 1.174
MDL Number: MFCD00153204
CAS Number: 13630-19-8
UN: 1993
ADR: 3 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty