Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Iodopropyl)trimethoxysilane 98%

Supplier: AMBEED, INC
Warning

A555215-100G A555215-25G A555215-5G
77157-184EA 188 USD InStock
77157-184 77157-186 77157-188
(3-Iodopropyl)trimethoxysilane 98%
(3-Iodopropyl)trimethoxysilane
Precaución: for Research Use Only


Formula: IH₂CCH₂CH₂Si(OCH₃)₃
MDL Number: MFCD00054838
CAS Number: 14867-28-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty