Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-([2,2'-Bipyridin]-5-yl)-2-aminopropanoic acid 95%

Supplier: AMBEED, INC
Warning

A567531-100MG A567531-250MG
77157-282EA 381 USD InStock
77157-282 77157-284
3-([2,2'-Bipyridin]-5-yl)-2-aminopropanoic acid 95%
3-([2,2'-Bipyridin]-5-yl)-2-aminopropanoic acid
Precaución: for Research Use Only


Formula: C₁₃H₁₃N₃O₂
MDL Number: MFCD28899022
CAS Number: 1219368-79-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty