Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-5-Benzyl-2,2,3-trimethylimidazolidin-4-one hydrochloride 95%

Supplier: AMBEED, INC
Warning

A573519-100MG A573519-1G A573519-250MG A573519-5G
77157-356EA 31 USD InStock
77157-356 77157-358 77157-360 77686-460
(R)-5-Benzyl-2,2,3-trimethylimidazolidin-4-one hydrochloride 95%
(R)-5-Benzyl-2,2,3-trimethylimidazolidin-4-one hydrochloride
Precaución: for Research Use Only


Formula: C₁₃H₁₉ClN₂O
MDL Number: MFCD08276839
CAS Number: 323196-43-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty