Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Chlorophenyl)benzimidazole 98%

Supplier: AMBEED, INC
Warning

A578674-1G A578674-250MG A578674-5G
77157-422EA 53 USD InStock
77157-422 77157-424 77157-426
2-(4-Chlorophenyl)benzimidazole 98%
2-(4-Chlorophenyl)benzimidazole
Precaución: for Research Use Only


Formula: C₁₃H₉ClN₂
MDL Number: MFCD00224359
CAS Number: 1019-85-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty