Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Methoxy-[1,1'-biphenyl]-4-amine 98%

Supplier: AMBEED, INC
Warning

A584218-1G A584218-250MG A584218-5G
77157-452EA 36 USD InStock
77157-452 77157-454 77157-456
4'-Methoxy-[1,1'-biphenyl]-4-amine 98%
4'-Methoxy-[1,1'-biphenyl]-4-amine
Precaución: for Research Use Only


Formula: C₁₃H₁₃NO
MDL Number: MFCD00047858
CAS Number: 1137-77-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty