Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

TTP-22 98%

Supplier: AMBEED, INC
Warning

A586763-100MG A586763-10MG A586763-1MG A586763-25MG A586763-50MG
77157-482EA 264.01 USD InStock
77157-482 77157-484 77157-486 77157-488 77157-490
TTP-22 98%
TTP-22
Precaución: for Research Use Only


Formula: C₁₆H₁₄N₂O₂S₂
MDL Number: MFCD02654497
CAS Number: 329907-28-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty