Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-5-bromopyrrolo[1,2-f][1,2,4]triazine 95%

Supplier: AMBEED, INC
Warning

A601836-100MG A601836-1G A601836-250MG
77157-590EA 65 USD InStock
77157-590 77157-592 77157-594
4-Chloro-5-bromopyrrolo[1,2-f][1,2,4]triazine 95%
4-Chloro-5-bromopyrrolo[1,2-f][1,2,4]triazine
Precaución: for Research Use Only


Formula: C₆H₃BrClN₃
MDL Number: MFCD23106343
CAS Number: 1403767-33-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty