Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Nitro-4-(trifluoromethyl)benzoyl chloride 95%

Supplier: AMBEED, INC
Danger

A605989-1G A605989-25G A605989-5G
77157-638EA 14 USD InStock
77157-638 77157-640 77157-642
2-Nitro-4-(trifluoromethyl)benzoyl chloride 95%
2-Nitro-4-(trifluoromethyl)benzoyl chloride
Precaución: for Research Use Only


Formula: C₈H₃ClF₃NO₃
MW: 253.57 g/mol
MDL Number: MFCD07777203
CAS Number: 81108-81-8
UN: 1760
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty