Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-1-(2-bromo-4-nitrophenyl)ethanone 95%

Supplier: AMBEED, INC
Danger

A625578-100MG A625578-1G A625578-250MG
77157-806EA 31 USD InStock
77157-806 77157-808 77157-810
2-Bromo-1-(2-bromo-4-nitrophenyl)ethanone 95%
2-Bromo-1-(2-bromo-4-nitrophenyl)ethanone
Precaución: for Research Use Only


Formula: C₈H₅Br₂NO₃
MDL Number: MFCD08753757
CAS Number: 928709-80-2
UN: 3261
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty