Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-D-tyrosine 98%

Supplier: AMBEED, INC
Warning

A629999-100G A629999-1G A629999-25G A629999-5G
77157-844EA 327 USD InStock
77157-844 77157-846 77157-848 77157-850
Fmoc-D-tyrosine 98%
Fmoc-D-tyrosine
Precaución: for Research Use Only


Formula: C₂₄H₂₁NO₅
MW: 403.434 g/mol
MDL Number: MFCD00171384
CAS Number: 112883-29-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty