Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

GNF-5837 98+%

Supplier: AMBEED, INC
Warning

A638163-10MG A638163-1MG A638163-50MG A638163-5MG
77157-902EA 65 USD InStock
77157-902 77157-904 77157-906 77157-908
GNF-5837 98+%
GNF-5837
Precaución: for Research Use Only


Formula: C₂₈H₂₁F₄N₅O₂
MW: 535.5 g/mol
MDL Number: MFCD24386809
CAS Number: 1033769-28-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty