Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Mal-amido-PEG6-NHS 95%

Supplier: AMBEED, INC
Warning

A642798-1MG A642798-100MG A642798-1G A642798-250MG
77157-968EA 10 USD InStock
77157-968 77686-591 77686-592 77686-593
Mal-amido-PEG6-NHS 95%
Mal-amido-PEG6-NHS
Precaución: for Research Use Only


Formula: C₂₆H₃₉N₃O₁₃
MDL Number: MFCD11041093
CAS Number: 1137109-21-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty