Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxy-3-methoxy-5-methylbenzaldehyde 95%

Supplier: AMBEED, INC
Warning

A648672-100MG A648672-1G A648672-250MG
77157-994EA 47 USD InStock
77157-994 77157-996 77157-998
2-Hydroxy-3-methoxy-5-methylbenzaldehyde 95%
2-Hydroxy-3-methoxy-5-methylbenzaldehyde
Precaución: for Research Use Only


Formula: C₉H₁₀O₃
MW: 166.17 g/mol
MDL Number: MFCD06740697
CAS Number: 7452-10-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty