Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-chloro-3-nitroisonicotinate 98%

Supplier: AMBEED, INC
Warning

A654745-100MG A654745-1G A654745-250MG A654745-5G
77158-010EA 20 USD InStock
77158-010 77158-012 77158-014 77158-016
Methyl 2-chloro-3-nitroisonicotinate 98%
Methyl 2-chloro-3-nitroisonicotinate
Precaución: for Research Use Only


Formula: C₇H₅ClN₂O₄
MDL Number: MFCD17214977
CAS Number: 1379302-09-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty