Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile 96%

Supplier: AMBEED, INC
Warning

A661352-100MG A661352-1G A661352-250MG
77158-068EA 37 USD InStock
77158-068 77158-070 77158-072
4-Amino-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile 96%
4-Amino-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
Precaución: for Research Use Only


Formula: C₁₃H₁₇BN₂O₂
MDL Number: MFCD12025487
CAS Number: 1315350-04-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty