Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 4-(3,3,3-trifluoro-2-hydroxypropyl)piperidine-1-carboxylate 95%

Supplier: AMBEED, INC
Warning

A662482-100MG A662482-1G A662482-250MG
77158-078EA 30 USD InStock
77158-078 77158-080 77158-082
tert-Butyl 4-(3,3,3-trifluoro-2-hydroxypropyl)piperidine-1-carboxylate 95%
tert-Butyl 4-(3,3,3-trifluoro-2-hydroxypropyl)piperidine-1-carboxylate
Precaución: for Research Use Only


Formula: C₁₃H₂₂F₃NO₃
MDL Number: MFCD16618527
CAS Number: 1228631-10-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty