Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Piperidinium-1-ylmethyl)trifluoroborate 98%

Supplier: AMBEED, INC
Danger

A665504-100MG A665504-1G A665504-250MG
77158-104EA 10 USD InStock
77158-104 77158-106 77158-108
(Piperidinium-1-ylmethyl)trifluoroborate 98%
(Piperidinium-1-ylmethyl)trifluoroborate
Precaución: for Research Use Only


Formula: C₆H₁₃BF₃N
MW: 166.98 g/mol
MDL Number: MFCD18071160
CAS Number: 1268340-93-7
UN: 1759
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty