Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-fluorobenzenesulphonyl chloride 97%

Supplier: AMBEED, INC
Danger

A674436-1G A674436-250MG A674436-5G A674436-25G
77158-182EA 6 USD InStock
77158-182 77158-184 77158-186 77686-639
2-Chloro-4-fluorobenzenesulphonyl chloride 97%
2-Chloro-4-fluorobenzenesulphonyl chloride
Precaución: for Research Use Only


Formula: C₆H₃Cl₂FO₂S
MW: 229.06 g/mol
MDL Number: MFCD00052957
CAS Number: 85958-57-2
UN: 3265
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty