Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl 3-bromophthalate 98%

Supplier: AMBEED, INC
Warning

A690537-1G A690537-250MG A690537-5G
77158-318EA 68 USD InStock
77158-318 77158-320 77158-322
Dimethyl 3-bromophthalate 98%
Dimethyl 3-bromophthalate
Precaución: for Research Use Only


Formula: C₁₀H₉BrO₄
MDL Number: MFCD10687082
CAS Number: 58749-33-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty