Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Catecholbis(trifluoromethanesulfonate) 98%

Supplier: AMBEED, INC
Danger

A693676-1G A693676-250MG A693676-5G
77158-368EA 46 USD InStock
77158-368 77158-370 77158-372
Catecholbis(trifluoromethanesulfonate) 98%
Catecholbis(trifluoromethanesulfonate)
Precaución: for Research Use Only


Formula: C₈H₄F₆O₆S₂
MDL Number: MFCD00274274
CAS Number: 17763-91-6
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty