Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

TTNPB 98%

Supplier: AMBEED, INC
Danger

A700949-10MG A700949-1MG A700949-50MG A700949-5MG
77158-426EA 40 USD InStock
77158-426 77158-428 77158-430 77158-432
TTNPB 98%
TTNPB
Precaución: for Research Use Only


Formula: C₂₄H₂₈O₂
MW: 348.5 g/mol
MDL Number: MFCD00673917
CAS Number: 71441-28-6
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty