Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-fluoro-5-iodobenzoate 98%

Supplier: AMBEED, INC
Warning

A703759-100MG A703759-1G A703759-250MG A703759-5G
77158-458EA 9 USD InStock
77158-458 77158-460 77158-462 77158-464
Methyl-2-fluoro-5-iodobenzoate 98%
Methyl 2-fluoro-5-iodobenzoate
Precaución: for Research Use Only


Formula: C₈H₆FIO₂
MW: 280.0349 g/mol
MDL Number: MFCD06203930
CAS Number: 625471-27-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty