Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium phenoxymethyltrifluoroborate 95%

Supplier: AMBEED, INC
Warning

A705293-250MG A705293-1G
77158-476EA 44 USD InStock
77158-476 77686-690
Potassium phenoxymethyltrifluoroborate 95%
Potassium phenoxymethyltrifluoroborate
Precaución: for Research Use Only


Formula: C₇H₇BF₃KO
MW: 214.04 g/mol
MDL Number: MFCD10566516
CAS Number: 1027642-30-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty