Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(4-Chloro-3-methoxyphenyl)picolinamide 98%

Supplier: AMBEED, INC
Warning

A709934-100MG A709934-10MG A709934-25MG A709934-50MG A709934-5MG
77158-516EA 32 USD InStock
77158-516 77158-518 77158-520 77158-522 77158-524 77158-526
N-(4-Chloro-3-methoxyphenyl)picolinamide 98%
N-(4-Chloro-3-methoxyphenyl)picolinamide
Precaución: for Research Use Only


Formula: C₁₃H₁₁ClN₂O₂
MDL Number: MFCD16618402
CAS Number: 1161205-04-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty