Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-nitrobenzophenone 98%

Supplier: AMBEED, INC
Warning

A714633-100G A714633-25G A714633-5G
77158-582EA 87 USD InStock
77158-582 77158-584 77158-586
2-Chloro-5-nitrobenzophenone 98%
2-Chloro-5-nitrobenzophenone
Precaución: for Research Use Only


Formula: C₁₃H₈ClNO₃
MW: 261.66 g/mol
MDL Number: MFCD00007295
CAS Number: 34052-37-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty