Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

MLN-4924 98%

Supplier: AMBEED, INC
Warning

A724332-1MG A724332-5MG
77158-630EA 44 USD InStock
77158-630 77158-632
MLN-4924 98%
MLN-4924
Precaución: for Research Use Only


Formula: C₂₁H₂₅N₅O₄S
MDL Number: MFCD17215201
CAS Number: 905579-51-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty