Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(+)-4-Phenyl-2-butanol 98%

Supplier: AMBEED, INC
Warning

A724453-100MG A724453-1G A724453-250MG
77158-634EA 12 USD InStock
77158-634 77158-636 77158-638
(S)-(+)-4-Phenyl-2-butanol 98%
(S)-(+)-4-Phenyl-2-butanol
Precaución: for Research Use Only


Formula: C₁₀H₁₄O
MDL Number: MFCD03093082
CAS Number: 22148-86-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty