Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Isopropylbenzaldehyde 97%

Supplier: AMBEED, INC
Warning

A726889-1000G A726889-100G A726889-25G A726889-500G
77158-670EA 149 USD InStock
77158-670 77158-672 77158-674 77158-676
4-Isopropylbenzaldehyde 97%
4-Isopropylbenzaldehyde
Precaución: for Research Use Only


Formula: C₁₀H₁₂O
MW: 148.2 g/mol
Boiling Pt: 235
MDL Number: MFCD00006953
CAS Number: 122-03-2
Merck Index: 13,02648
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty