Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2-Dioxido-1,2,3-oxathiazolidine, N-BOC protected 98%

Supplier: AMBEED, INC
Warning

A733633-100G A733633-10G A733633-1G A733633-25G A733633-5G
77158-704EA 427 USD InStock
77158-704 77158-706 77158-708 77158-710 77158-712
2,2-Dioxido-1,2,3-oxathiazolidine, N-BOC protected 98%
2,2-Dioxido-1,2,3-oxathiazolidine, N-BOC protected
Precaución: for Research Use Only


Formula: C₇H₁₃NO₅S
MDL Number: MFCD20527196
CAS Number: 459817-82-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty