Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(4-Aminophenyl)-1,3,4-oxadiazole-2-thiol 95%

Supplier: AMBEED, INC
Warning

A734143-25G A734143-5G
77158-724EA 220 USD InStock
77158-724 77158-726
5-(4-Aminophenyl)-1,3,4-oxadiazole-2-thiol 95%
5-(4-Aminophenyl)-1,3,4-oxadiazole-2-thiol


Formula: C₈H₇N₃OS
MW: 193.23 g/mol
MDL Number: MFCD00806252
CAS Number: 32058-82-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty