Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-chloro-5-methoxypicolinate 95%

Supplier: AMBEED, INC
Warning

A741777-100MG A741777-250MG
77158-800EA 39 USD InStock
77158-800 77158-802
Methyl 4-chloro-5-methoxypicolinate 95%
Methyl 4-chloro-5-methoxypicolinate
Precaución: for Research Use Only


Formula: C₈H₈ClNO₃
MDL Number: MFCD18257711
CAS Number: 857433-67-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty