Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(+)-Clopidogrel sulfate 98%

Supplier: AMBEED, INC
Warning

A749538-100G A749538-100MG A749538-1MG A749538-25G A749538-25MG A749538-5G
77158-854EA 340.99 USD InStock
77158-854 77158-856 77158-858 77158-860 77158-862 77158-864
(S)-(+)-Clopidogrel sulfate 98%
(S)-(+)-Clopidogrel sulfate
Precaución: for Research Use Only


Formula: C₁₆H₁₆ClNO₂S
MDL Number: MFCD00876395
CAS Number: 120202-66-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty