Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-chlorobenzenesulfonyl chloride 96%

Supplier: AMBEED, INC
Danger

A760806-25G A760806-5G
77158-956EA 44 USD InStock
77158-956 77158-958
4-Bromo-2-chlorobenzenesulfonyl chloride 96%
4-Bromo-2-chlorobenzenesulphonyl chloride
Precaución: for Research Use Only


Formula: C₆H₃BrCl₂O₂S
MDL Number: MFCD03094630
CAS Number: 351003-52-6
UN: 3261
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty