Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-tert-Butyl 2-methyl 3,3-dimethyl-4-oxopyrrolidine-1,2-dicarboxylate 95%

Supplier: AMBEED, INC
Warning

A768373-100MG A768373-1G A768373-250MG
77158-992EA 98 USD InStock
77158-992 77158-994 77158-996
1-tert-Butyl 2-methyl 3,3-dimethyl-4-oxopyrrolidine-1,2-dicarboxylate 95%
1-tert-Butyl 2-methyl 3,3-dimethyl-4-oxopyrrolidine-1,2-dicarboxylate
Precaución: for Research Use Only


Formula: C₁₃H₂₁NO₅
MW: 271.31 g/mol
MDL Number: MFCD24470049
CAS Number: 848444-88-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty